Order Entry
United Kingdom
ContactUsLinkComponent
 
Pesticide reference standard, 2,3-Dichloroaniline 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA12322300ME
 :  
Pesticide reference standard, 2,3-Dichloroaniline 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA12322300ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    608-27-5
  • Description:
    2,3-Dichloroaniline 100 µg/ml in methanol
  • Formula:
    C₆H₅Cl₂N
  • Description:
    2,3-Dichloroaniline