Order Entry
United Kingdom
ContactUsLinkComponent
Ethylene carbonate
Ethylene carbonate
Catalog # PROONIST-1972
CAS Number:  
Ethylene carbonate
Catalog # PROONIST-1972
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    60 g
  • Seq:
    3377

Specifications

About this item

 :
 :88.06 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:36 °C
Density:1.47 g/cm³ (20 °C)
Flash Pt:150 °C
Storage Temperature:Ambient
EINECS:202-510-0
 :GHS05
CAS Number:96-49-1
Formula:C₃H₄O₃
MDL Number:MFCD00005382