Order Entry
United Kingdom
ContactUsLinkComponent
Pesticide reference standard, 3,5-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 3,5-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12453500ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 3,5-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12453500ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    591-35-5
  • Description:
    3,5-Dichlorophenol 100 µg/ml in methanol
  • Formula:
    C₆H₄Cl₂O
  • Description:
    3,5-Dichlorophenol

Specifications

About this item