Pesticide reference standard, 2,4-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA12451000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:120-83-2
- Description:2,4-Dichlorophenol 100 µg/ml in methanol
- Formula:C₆H₄Cl₂O
- Description:2,4-Dichlorophenol