

2-(Trifluoromethoxy)phenylacetonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:137218-25-8
:
Formula:C₉H₆F₃NO
MDL Number:MFCD00236326
:201.15
Boiling Pt:222…223 °C
Density:1.280
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together