1-(4-Chlorophenyl)piperazine
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:38212-33-8
:
Formula:C₁₀H₁₃ClN₂
MDL Number:MFCD00044823
:196.68 g/mol
Boiling Pt:137…138,5 °C (1 mmHg)
Melting Pt:71,5…73.5 °C
Frequently Bought Together