Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
1,3-Dibromopropane

1,3-Dibromopropane

Flammable
Irritant
Environment
Marketsource
Supplier:   FLUOROCHEM
1,3-Dibromopropane
Supplier:  FLUOROCHEM
Marketsource
Certificates
1,3-Dibromopropane

2 Options Available Below

Product Details & Documents
  • CAS Number:109-64-8
  • Formula:BrH₂CCH₂CH₂Br
  • MDL Number:MFCD00000255
  • Molecular Weight:201.89 g/mol
  • Merck Index:12,09844
  • Boiling Pt:167 °C (1013 hPa)
  • Melting Pt:–34 °C
  • Density:1.99 g/cm³ (20 °C)
  • Flash Pt:47 °C
  • Storage Temperature:Ambient
  • EINECS:203-690-3
  • UN:1993
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource