1,12-Dibromododecane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3344-70-5
:
Formula:BrH₂C(CH₂)₁₀CH₂Br
MDL Number:MFCD00000226
:328.13 g/mol
Boiling Pt:215 °C (15 mmHg)
Melting Pt:38…42 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:222-096-5
Related Information
Frequently Bought Together