
1,1,2,2-Tetrabromoethane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-27-6
:
Formula:Br₂HCCHBr₂
MDL Number:MFCD00000133
:345.65 g/mol
Merck Index:13,09261
Boiling Pt:244 °C (1013 hPa)
Melting Pt:1 °C
Density:2.96 g/cm³
Storage Temperature:Ambient
EINECS:201-191-5
UN:2504
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.6355 to 1.6375 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤75 APHA
Specification Test Results
Frequently Bought Together