Sodium molybdate(VI) dihydrate, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
CAS Number:10102-40-6
:
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170
Density:3.28 g/cm³ (20 °C)
Molecular Weight:241.95 g/mol
Melting Pt:687
Merck Index:13,08718
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-551-7
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:840 g/l (20 °C)
- Melting Point:687 °C Elimination of water of crystallisation
- Molar Mass:241.95 g/mol
- Bulk Density:1000 - 1400 kg/m3
- pH:9 - 10 (840 g/l, H2O, 20 °C)
- Hill Formula:Na2MoO4 * 2 H2O
- Chemical Formula:Na2MoO4 * 2 H2O
Specification Test Results
Related Information
Frequently Bought Together