Sodium hydrogen carbonate 75 g/l in aqueous solution
Catalog # ALPDBICA0002012
Supplier: Alpha Diagnostics International
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:84.01 g/mol
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528