


Trimethylamine 45% (w/w) in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-50-3
:
Formula:C₃H₉N
MDL Number:MFCD00008327
Boiling Pt:30 °C (1013 hPa)
Melting Pt:–3 °C
Density:0.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-875-0
UN:1297
ADR:3,I
:59.11 g/mol
:GHS02
:GHS05
:GHS07
Frequently Bought Together