Hydrogen peroxide 35% (w/w) in aqueous solution stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:107 °C
Melting Pt:–26 °C
Density:1.130
Molecular Weight:34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
ADR:5.1 II
Merck Index:14,4798
UN:2014
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Color):Clear, colorless
- Form:Liquid
- Assay (Redox Titration):33,5 - 36,5%
- Comment::Stabilised with <0,1% w/v Proprietary Inhibitor
Specification Test Results
Related Information
Frequently Bought Together