Chromium, powder, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-47-3
:
Formula:Cr
MDL Number:MFCD00010944
ADR:4.1,III
:M-Clarity™ quality level = MQ100
:52 g/mol
EINECS:231-157-5
UN:3089
Frequently Bought Together