Order Entry
United Kingdom
ContactUsLinkComponent
Chromium, Patinal®
Chromium, Patinal®
Catalog # 1.11807.0100
Supplier:  Merck
CAS Number:  
Chromium, Patinal®
Catalog # 1.11807.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic can
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Merck Index:12,02288
Boiling Pt:2469 °C (1013 hPa)
Melting Pt:1840 °C
Density:7.2 g/cm³ (20 °C)
 :52 g/mol
Storage Temperature:Ambient
EINECS:231-157-5
CAS Number:7440-47-3
Formula:Cr
MDL Number:MFCD00010944