Magnetic stirring bars, PTFE, star-shaped, Spinstar®
: Brand
Triangular shaped PTFE magnetic stirring bars.
- Ideal for dissolving solids and mixing sediments due to scraping action
- Angular surface creates high turbulence even at low speeds
Frequently Bought Together