
Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7723-14-0
:
Formula:P
MDL Number:MFCD00133771
:30.97 g/mol
Density:∼2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-768-7
UN:1338
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together