Antimony 99+%, powder 0.15 mm
Catalog # ACRO192450100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Grey Powder
- Assay:≥99.0 %
Specification Test Results
About this item
:
:121.76 g/mol
Melting Pt:630 °C
Storage Temperature:Ambient
EINECS:231-146-5
UN:2871
ADR:6.1,III
:GHS08
:GHS09
CAS Number:7440-36-0
Formula:Sb
MDL Number:MFCD00134030