Order Entry
United Kingdom
ContactUsLinkComponent
Calcium 99%, pieces
Calcium 99%, pieces
Certificates
undefined
Calcium 99%, pieces

Specification Test Results

  • Appearance:
    Grey Metallic pieces
  • Assay:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together