

Lithium, dry 99+%, granular
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-93-2
:
Formula:Li
MDL Number:MFCD00134051
:6.94 g/mol
Merck Index:13,05542
Boiling Pt:1317 °C (1013 hPa)
Melting Pt:180 °C
Density:0,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-102-5
UN:1415
ADR:4.3,I
:GHS02
:GHS05
Specification Test Results
- Appearance:Silver grey Granulate
- Assay:≥99.0 %
Specification Test Results
Frequently Bought Together