Potassium chloride 3.3 mol/l (3.3 N) in aqueous solution
PRLSMC311264.2000
:
- Pack type:Plastic bottle
- Pk:2 L
- Seq:5086
- Environmentally Preferable:
: Potassiumchloride
:
:74.55 g/mol
:1420
:773
:1.13 g/cm³ (20 °C)
:Ambient
:231-211-8
:7447-40-7
:KCl
:MFCD00011360
:14,07621