2,2',4,4'-Tetrahydroxybenzophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:131-55-5
:
Formula:C₁₃H₁₀O₅
MDL Number:MFCD00002278
:246.22 g/mol
Boiling Pt:100…102 °C (40 mmHg)
Melting Pt:196…198 °C
Density:1.498 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:205-028-9
Frequently Bought Together