Order Entry
United Kingdom
ContactUsLinkComponent
 
Bovine serum albumin (BSA), Fraction V
 :  Serva
 
Bovine serum albumin (BSA), Fraction V
 :  Serva

 

  • CAS No.:
    90604-29-8
    9048-46-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • pH:
    pH 7
  • Grade:
    Standard Grade

 

Frequently Bought Together