Order Entry
United Kingdom
ContactUsLinkComponent
Reverse osmosis systems, RO 10 / 20 / 30 / 40
Reverse osmosis systems, RO 10 / 20 / 30 / 40
Certificates
undefined
Reverse osmosis systems, RO 10 / 20 / 30 / 40
Specifications
  • Catalog No:
  • 172-4213
  • 172-4212
  • 172-4211
  • 172-4214
  • Model:
  • RO 30
  • RO 20
  • RO 10
  • RO 40
  • Feed water:
  • Tap water (according to WHO)
  • Tap water (according to WHO)
  • Tap water (according to WHO)
  • Tap water (according to WHO)
  • Production flow rate:
  • 30 L/h
  • 20 L/h
  • 10 L/h
  • 40 L/h
  • W×D×H:
  • 500×295×600 mm
  • 500×295×600 mm
  • 500×295×600 mm
  • 500×295×600 mm
  • Weight:
  • 32 kg
  • 30 kg
  • 30 kg
  • 32 kg