Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sponge, reusable
Sponge, reusable
Yellow, 140×115 mm
Catalog # 129-0920
Supplier:   DISTRIBUTOR
Sponge, reusable
Catalog # 129-0920
Supplier:  DISTRIBUTOR
Sponge, reusable
Sponge, reusable
Yellow, 140×115 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Durable sponges suitable for the cleaning of tough stains and wiping of oil spillages.
  • Cellulose toughened with fibre, provides more absorbency
  • Not adversely affected by oil, grease or petrol
Specifications
  • Colour:
    Yellow
  • L×W:
    140×115 mm