Viscosity cups with nozzle
Anodised aluminium and stainless steel viscometer cups with nozzle for break-thread flow time comparative measurements.
- Easy cleaning
- Engraved with a unique serial number
Typically used for the consistency of paintings, varnishes and similar products.
: DIN 53211
: The stand should be ordered separately.
Frequently Bought Together