U-tube viscometers
The time for the level of the liquid to pass between these marks is proportional to the kinematic viscosity.
- Upper bulb and bulb lower down on the other arm
- Two marks (one above and one below the upper bulb) indicate a known volume
Liquid is drawn into the upper bulb by suction, then flowed down through the capillary into the lower bulb.
: UKAS certified. ISO 3104, 3105, ASTM D445/446, IP 71, BS 188
- Catalog No:
- 616-3311
- 616-3310
- 616-3313
- 616-3308
- 616-3312
- 616-3315
- 616-3314
- 616-3309
- Length:
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- Volume:
- 13 − 40 ml
- 13 − 40 ml
- 13 − 40 ml
- 13 − 40 ml
- 13 − 40 ml
- 13 − 40 ml
- 13 − 40 ml
- 13 − 40 ml
Frequently Bought Together