Test tubes, reductase
Supplier: JONES FR
Jump to:
About this item
Heavy wall rimless soda glass test tubes.
- Yellow notation patch
- Ceramic enamel ring marking at appropriate capacity
4 Options Available Below
Product Details & Documents
Heavy wall rimless soda glass test tubes.
- Yellow notation patch
- Ceramic enamel ring marking at appropriate capacity
Product Family Options
Product Information
- AvailabilityPrice
- 212-2201Tubes with marks at 2,5 and 10 ml for antibiotic tests
- 212-2202Tubes with marks at 5 and 10 ml for Phosphatase and COD testsTubes with marks at 5 and 10 ml for Phosphatase and COD testsLog in for availability1 * 100 items£180.00
- 212-2203Tubes with marks at 7 and 10 ml for ice cream tests
- 212-2200Tubes with marks at 10 ml for methylene blue reduction testTubes with marks at 10 ml for methylene blue reduction testLog in for availability1 * 100 items£180.00