Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Labelling tapes, temperature resistant

Labelling tapes, temperature resistant

Labelling tapes, temperature resistant
Certificates
Labelling tapes, temperature resistant
Labelling tapes, temperature resistant

173 Options Available Below

Product Details & Documents

This all-purpose, self-adhesive tape is well suited for a wide variety of labelling applications. It can be written on with pen, pencil or permanent marker.

  • Temperature resistant from −45 to +121 °C
  • With removable adhesive
  • Oil, water, and acid resistant, but not designed to be fully submerged in water

This tape is available in various colours, in two lengths and five widths.

Product Family Options

Length 12,7 m and core 25 mm

Product Information

  • Colour
    Width
    Availability
    Price
  • 817-1501
    Tape, temperature resistant
    Tape, temperature resistant
    White
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1502
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1503
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1504
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1505
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1506
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1507
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1508
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1509
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1510
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1511
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1512
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1513
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1514
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-0039
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-0040
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-0041
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1515
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £10.80
    £10.80
    1 * 1 Roll
       £10.80

  • 817-1521
    Tape, temperature resistant
    Tape, temperature resistant
    White
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1522
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1523
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1524
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1525
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1526
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1527
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1528
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1529
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1530
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1531
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1532
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1533
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1534
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-0373
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-0043
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-0374
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1535
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £14.40
    £14.40
    1 * 1 Roll
       £14.40

  • 817-1541
    Tape, temperature resistant
    Tape, temperature resistant
    White
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1542
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1543
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1544
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1545
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1546
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1547
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1548
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1549
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1550
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1551
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1552
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1553
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1554
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1555
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £18.10
    £18.10
    1 * 1 Roll
       £18.10

  • 817-1538
    Tape, temperature resistant
    Tape, temperature resistant
    White
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-1539
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0368
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0369
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0370
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-1540
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-1556
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0371
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0359
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0360
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0361
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0362
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0363
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0364
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0365
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0366
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • 817-0367
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £50.40
    £50.40
    1 * 1 Roll
       £50.40

  • TIMET-502-1
    Tape, temperature resistant
    Tape, temperature resistant
    White
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-2
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-3
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-4
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-5
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-6
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-7
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-8
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-9
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-10
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-11
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-12
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-13
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-14
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-15
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-16
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-17
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

  • TIMET-502-18
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £59.10
    £59.10
    1 * 1 Roll
       £59.10

Length 54,9 m and core 76 mm

Product Information

  • Colour
    Width
    Availability
    Price
  • 817-1600
    Tape, temperature resistant
    Tape, temperature resistant
    White
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1601
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1602
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1603
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1604
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1605
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1606
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1607
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1608
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1609
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1610
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1611
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1612
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1613
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-0045
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-0046
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-0047
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1614
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £27.60
    £27.60
    1 * 1 Roll
       £27.60

  • 817-1615
    Tape, temperature resistant
    Tape, temperature resistant
    White
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1616
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1617
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1618
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1619
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1620
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1621
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1622
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1623
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1624
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1625
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1626
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1627
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1628
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-0049
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-0050
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1629
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £38.20
    £38.20
    1 * 1 Roll
       £38.20

  • 817-1630
    Tape, temperature resistant
    Tape, temperature resistant
    White
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1631
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1632
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1633
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1634
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1635
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1636
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1637
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1638
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1639
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1640
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1641
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1642
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1643
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-0218
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-0048
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-1644
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £49.80
    £49.80
    1 * 1 Roll
       £49.80

  • 817-0313
    Tape, temperature resistant
    Tape, temperature resistant
    White
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0323
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0324
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0325
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0326
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0327
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0328
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0329
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0330
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0315
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0316
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0317
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0318
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0319
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0320
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0321
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0322
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £69.00
    £69.00
    1 * 1 Roll
       £69.00

  • 817-0035
    Tape, temperature resistant
    Tape, temperature resistant
    White
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0021
    Tape, temperature resistant
    Tape, temperature resistant
    Yellow
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0022
    Tape, temperature resistant
    Tape, temperature resistant
    Green
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0023
    Tape, temperature resistant
    Tape, temperature resistant
    Pink
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0024
    Tape, temperature resistant
    Tape, temperature resistant
    Red
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-6
    Tape, temperature resistant
    Tape, temperature resistant
    Orange
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0025
    Tape, temperature resistant
    Tape, temperature resistant
    Blue
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-8
    Tape, temperature resistant
    Tape, temperature resistant
    Tan
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-9
    Tape, temperature resistant
    Tape, temperature resistant
    Salmon
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-10
    Tape, temperature resistant
    Tape, temperature resistant
    Copper
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0036
    Tape, temperature resistant
    Tape, temperature resistant
    Rose
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-12
    Tape, temperature resistant
    Tape, temperature resistant
    Violet
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-13
    Tape, temperature resistant
    Tape, temperature resistant
    Pale purple
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-14
    Tape, temperature resistant
    Tape, temperature resistant
    Grey
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • 817-0037
    Tape, temperature resistant
    Tape, temperature resistant
    Light green
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-16
    Tape, temperature resistant
    Tape, temperature resistant
    Turquoise
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-17
    Tape, temperature resistant
    Tape, temperature resistant
    Green/yellow
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

  • TIMET-260-18
    Tape, temperature resistant
    Tape, temperature resistant
    Black
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - £79.60
    £79.60
    1 * 1 Roll
       £79.60

Complementary Products

Product Information

  • Catalog #
    For Use With
    Description
    W×D×H
    Availability
    Price
  • 817-0312
    Tapes with 28 mm width
    Handheld tape dispenser, HD-2, for rolls with up to 28 mm cores
    Log in for availability
    1 * 1 items - £17.90

  • 817-1592
    Tapes with 25 mm width
    Multiple roll dispenser, 500P
    250×114×86 mm
    Log in for availability
    1 * 1 items - £159.00

  • 817-1645
    Multiple roll dispenser, 60P, for rolls with 76 mm cores
    250×184×152 mm
    Log in for availability
    1 * 1 items - £286.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.