Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Stand plates

Stand plates

250 mm, 2,00 kg, 160 mm
Catalog # 241-0181
Supplier:   R L ENTERPRISES
Stand plates
Catalog # 241-0181
Supplier:  R L ENTERPRISES
Stand plates
Stand plates

250 mm, 2,00 kg, 160 mm

Price
£74.50
Log in for availability
1 * 1 items
Product Details & Documents

Steel, with blue acrylic finish. Rectangular stand base with 10 mm diameter threaded bore, 1,5 mm deep on side.

  • Resistant, radiant blue surface
  • Easy to clean
  • Anti-skid rubber feet for a firm footing
Specifications
  • Width:
    160 mm
  • Length:
    250 mm
  • Weight:
    2,00 kg