Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Stand plates

Stand plates

315 mm, 3,00 kg, 200 mm
Catalog # 241-0185
Supplier:   R L ENTERPRISES
Stand plates
Catalog # 241-0185
Supplier:  R L ENTERPRISES
Stand plates
Stand plates

315 mm, 3,00 kg, 200 mm

Price
£99.10
Log in for availability
1 * 1 items
Product Details & Documents

Steel, with blue acrylic finish. Rectangular stand base with 10 mm diameter threaded bore, 1,5 mm deep in the middle of the plate.

  • Resistant, radiant blue surface
  • Easy to clean
  • Anti-skid rubber feet for a firm footing
Specifications
  • Width:
    200 mm
  • Length:
    315 mm
  • Weight:
    3,00 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.