Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Transfer pipettes
Transfer pipettes
Supplier:   MEDLINE SCIENTIFIC
Transfer pipettes
Supplier:  MEDLINE SCIENTIFIC
Certificates
Transfer pipettes
Transfer pipettes
10 Options Available Below
Product Details & Documents
PE, disposable.
  • Extra long or graduated
  • Sterile models are ethylene oxide sterilised
Product Family Options
Product Information
  • Capacity
    Stem Ø
    Length
    Sterile
    Availability
    Price
  • 613-4981
    Transfer pipette, graduated (0,5 ml)
    Transfer pipette, graduated (0,5 ml)
    5 ml
    7,0 mm
    150 mm
    -
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £86.70
    £86.70
    1 * 500 items
       £86.70
  • 613-4982
    Transfer pipette, graduated (0,25 ml)
    Transfer pipette, graduated (0,25 ml)
    5 ml
    5,0 mm
    154
    -
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £22.70
    £22.70
    1 * 500 items
       £22.70
  • 613-4975
    Transfer pipette, graduated (0,25 ml)
    Transfer pipette, graduated (0,25 ml)
    5 ml
    5,0 mm
    154 mm
    +
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £64.70
    £64.70
    1 * 500 items
       £64.70
  • 613-4986
    Transfer pipette, graduated (each 0,25 ml)
    Transfer pipette, graduated (each 0,25 ml)
    1 ml
    5,0 mm
    154 mm
    +
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £179.00
    £179.00
    1 * 500 items
       £179.00
  • 613-4977
    Transfer pipette, graduated (each ml)
    Transfer pipette, graduated (each ml)
    5 ml
    - mm
    216 mm
    -
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - £51.30
    £51.30
    1 * 250 items
       £51.30
  • 613-4992
    Transfer pipette, not graduated
    Transfer pipette, not graduated
    6 ml
    225 mm
    -
    Log in for availability
    Log in for availability
    1 * 400 items
    1 * 400 items - £64.00
    £64.00
    1 * 400 items
       £64.00
  • 613-4984
    Transfer pipette, graduated (0,5 ml), individually wrapped
    Transfer pipette, graduated (0,5 ml), individually wrapped
    7 ml
    7,8 mm
    154 mm
    -
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £22.70
    £22.70
    1 * 500 items
       £22.70
  • 613-4985
    Transfer pipette, graduated (0,5 ml), individually wrapped
    Transfer pipette, graduated (0,5 ml), individually wrapped
    7 ml
    7,8 mm
    154 mm
    +
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £230.00
    £230.00
    1 * 500 items
       £230.00
  • 613-4988
    Transfer pipette, graduated (0,5 ml)
    Transfer pipette, graduated (0,5 ml)
    7 ml
    7,8 mm
    154 mm
    +
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - £150.00
    £150.00
    1 * 500 items
       £150.00
  • 613-4987
    Transfer pipette, not graduated
    Transfer pipette, not graduated
    23 ml
    300 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - £136.00
    £136.00
    1 * 100 items
       £136.00