Mounting medium
Mounting media, CRYO-M-BED, bottle of 113 ml, 1
Catalog # 720-0094
Supplier: BRIGHT INSTRUMENT
Jump to:
Product Details & Documents
CRYO-M-BED is virtually impossible to remove whilst frozen. Viscous enough to prevent thawing caused by capillary action creep up the sides of the block. It holds on to the specimen and the chuck firmly to allow consistent even sectioning. CYO-M-BED becomes opaque upon freezing, facilitating block insertion - late enough to minimise thawing and early enough for it to be held securely.
- Chemically inert
- Sectioning frozen CRYO-M-BED will not dull knife
- Miscible with water - facilitating easy removal
- High viscosity, strong adhesion to tissue and holder, enables specimen orientation while liquid
Specifications
Specifications
- Pk:1
- Description:Mounting media, CRYO-M-BED, bottle of 113 ml