Hand protectors, Hot Hand®
These gripping mittens made from flexible silicone elastomer can be used to handle hot or cold objects and to carry them short distances. The shape only protects the inside of the hand ensuring good sensitivity. The thumb must be inserted in one of the two openings, the other fingers in the remaining opening for a good grasping effect.
- Temperature range: −60 to 230 °C
- Clean with soapy water and store at room temperature away from damp and light
- Assist with grip and ensure good contact
Colour: White, red
EN 407, EN 511; Cat. II
: EN 407, EN 511; Cat. II
Frequently Bought Together