Nitrile gloves, TouchNTuff® 92-600 / 92-605
: Ansell
New Product
These gloves with rolled cuffs are made of puncture-resistant nitrile, which offers valuable protection against accidental splashes of hazardous chemicals.
- Non sterile, powder-free, smooth finish or textured fingertips
- 100% nitrile; no waxes, silicone, or plasticisers
- Ideal for sensitive skin
- Three times the puncture resistance of latex or PVC
- Light, cool, and durable
- Designed for maximum freedom of movement and minimum hand fatigue
Ideal for use in the chemical and electronics industry.
Thickness: 0,12 mm
Colour: Green
EN 374, EN 1149-5 (tested for EN 1149-1 and 1149-3), Cat. III; AQL 1.5
Food contact approved
: 92-600: EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5 (tested for EN 1149-1 and 1149-3), Cat. III; AQL 1,5
92-605: EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 421, EN 1149-5 (tested for EN 1149-1 and 1149-3), Cat. III; AQL 1,5
Food contact approved
92-605: EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 421, EN 1149-5 (tested for EN 1149-1 and 1149-3), Cat. III; AQL 1,5
Food contact approved
- Catalog No:
- 112-1937
- 112-6640
- 112-1936
- 112-6641
- 112-1938
- 112-0999
- 112-0997
- 112-0998
- 112-1935
- 112-1008
- Model:
- 92-605
- 92-600
- 92-605
- 92-600
- 92-605
- 92-600
- 92-600
- 92-600
- 92-605
- 92-600
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Smooth
- Textured fingertips
- Smooth
- Textured fingertips
- Smooth
- Smooth
- Smooth
- Textured fingertips
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 421, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 421, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 421, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 421, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
Frequently Bought Together