Gas sample tubes
Supplier: DIXON SCIENTIFIC
Jump to:
About this item
Borosilicate glass, grade 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Two capillary one-way stopcocks or two capillary three-way stopcocks with threaded stopcock arrester.
3 Options Available Below
Product Details & Documents
Borosilicate glass, grade 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Two capillary one-way stopcocks or two capillary three-way stopcocks with threaded stopcock arrester.
Product Family Options
Product Information