Triacetoxyvinylsilane, Sigma-Aldrich®
Catalog # SIAL483133-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:232.27 g/mol
EINECS:223-943-1
CAS Number:4130-08-9
Formula:C₈H₁₂O₆Si
MDL Number:MFCD00014975