Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Phthalide ≥98%

Phthalide ≥98%

Phthalide ≥98%
Certificates
Phthalide ≥98%
Phthalide ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 87-41-2 Molecular Formula: C8H6O2 Formula Weight: 134.13 Flash Point: 152°(305°F) Melting Point: 72-75° Boiling Point: 289-291° Beilstein Reference: 114632 MDL No.: MFCD00005906

Synonyms:  Isobenzofuran-1(3H)-one
  • CAS Number:87-41-2
  • Formula:C₈H₆O₂
  • MDL Number:MFCD00005906
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:289…291 °C
  • Melting Pt:72…75 °C
  • Density:1.1
  • Flash Pt:152 °C (305 °F)
  • Storage Temperature:Ambient
  • EINECS:201-744-0
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.