Order Entry
United Kingdom
ContactUsLinkComponent
Methyl cellulose, 450 mPa.S (2% H₂O) Reag. Ph. Eur., Sigma-Aldrich®
Methyl cellulose, 450 mPa.S (2% H₂O) Reag. Ph. Eur., Sigma-Aldrich®
Catalog # SIAL64632-500G
Supplier:  Merck
CAS Number:  
Methyl cellulose, 450 mPa.S (2% H₂O) Reag. Ph. Eur., Sigma-Aldrich®
Catalog # SIAL64632-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:9004-67-5
MDL Number:MFCD00081763