QuikPrep® Activated Charcoal Spin Columns
Catalog # HARV74-4806
Supplier: HARVARD APPARATUS
New Product
Specifications
- Description:Micro spin columns, activated charcoal, Elution Volume: 50 µl, Includes: Two 2 ml centrifuge tubes with top caps
- Seq:01
- Pk:24
- Sample volume:0,025 - 0,075 ml
Specifications
About this item
QuikPrep® SpinColumns™ with activated charcoal packing materials can be used for DNA, protein and peptide purification, small molecule, carbohydrate, salt and radiolabel removal, nick translation and affinity separation. Activated charcoal retains polar solutes. Salt will not be retained.