

Formaldehyde 0.4% in sulphuric acid 95% Reag. Ph. Eur. 1086805
Catalog # 87951.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:1830
ADR:8,II
:GHS05
:GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274