Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sodium azide 5% in aqueous solution

Sodium azide 5% in aqueous solution

Glass bottle, 500 mL
Catalog # PRLS88024.260
Toxic
Environment
Sodium azide 5% in aqueous solution
Catalog # PRLS88024.260
Supplier:  
CAS Number:  26628-22-8
Sodium azide 5% in aqueous solution

Glass bottle, 500 mL

CAS Number:  26628-22-8
Discontinued
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • EINECS:247-852-1
  • UN:3287
  • ADR:6.1,II

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications