Order Entry
United Kingdom
ContactUsLinkComponent
Sulphanilic acid 0.02 mol/l in acetic acid (6%) Reag. Ph. Eur. 1086201
Sulphanilic acid 0.02 mol/l in acetic acid (6%) Reag. Ph. Eur. 1086201
Catalog # 87944.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphanilic acid 0.02 mol/l in acetic acid (6%) Reag. Ph. Eur. 1086201
Catalog # 87944.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :173.19 g/mol
EINECS:204-482-5
CAS Number:121-57-3
Formula:4-(H₂N)C₆H₄SO₃H
MDL Number:MFCD00007886