Gel-Loading Pipette Tips
COLE25713-12
New Product
- Colour code:Natural
- Packed:5 packs, 204/pack
- Sterile:-
- Volume:1 - 200 µl
- Length:69,85 mm
- Pk:1
These extended microcapillary tips are excellent for loading electrophoresis gels. The polypropylene tips are rounded to fit easily into small instrument openings. Use these microcapillary tips for DNA sequencing, loading SDS PAGE gels, and other critical sampling applications.
Compatible with Biohit Proline, Costar®, Eppendorf®, Finnpipette®, Jencons Sealpette, Oxford® BenchMate™, Pipetman®, Socorex, Titertek® Digital.