Tiliroside 98%
Catalog # 471721000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:yellow powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:594.525 g/mol
Boiling Pt:944°C (760mmHg)
Flash Pt:311°C
Storage Temperature:Ambient
CAS Number:20316-62-5
Formula:C₃₀H₂₆O₁₃
MDL Number:MFCD00017454