Resiquimod 98%
Catalog # 471212500.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:white to yellow to beige crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:190°C to 195°C
- Water:≤2 %
Specification Test Results
About this item
:
:314.389 g/mol
Storage Temperature:Freezer
CAS Number:144875-48-9
Formula:C₁₇H₂₂N₄O₂
MDL Number:MFCD00937759