
Prulifloxacin 97%
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:123447-62-1
:
Formula:C₂₁H₂₀FN₃O₆S
MDL Number:MFCD00864847
:461.46 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:yellow to light orange to green powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together