Atomoxetine Hydrochloride
Glass bottle, 1 g
Catalog # 467680010.
Supplier: Thermo Fisher Scientific
Atomoxetine Hydrochloride
Catalog # 467680010.
Supplier: Thermo Fisher Scientific
CAS Number: 82248-59-7
Jump to:
Product Details & Documents
Synonyms: (R)-N-Methyl-3-(2-methylphenoxy)-3-phenylpropylamine Hydrochloride, (R)-Tomoxetine Hydrochloride, (R)-N-Methyl-3-phenyl-3-(o-tolyloxy)propan-1-amine hydrochloride
- CAS Number:82248-59-7
- Formula:C₁₇H₂₁NO
- MDL Number:MFCD06410992
- Molecular Weight:291.82 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:white to beige powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Melting point:167°C to 171°C
- Loss on drying:≤0.5 % (1 g, 105°C, 2 hr)
Specifications
- Pack type:Glass bottle
- Pk:1 g



