Eppendorf™ Microloader™ Pipette Tips
0,5 - 20 µl, Eppendorf™ microinjection capillaries
Catalog # DIUK10289651
Supplier: DISTRIBUTOR
Jump to:
Product Details & Documents
Use these extremely long, fine and flexible tips for filling of microcapillaries, racked. Eppendorf ™ Microloader™ pipette tips are ideal for micro-injection and where additional reach is needed.
Specifications
- Volume:0,5 - 20 µl
- For use with:Eppendorf™ microinjection capillaries