
Rhodium(II) triphenylacetate dimer (CH₂Cl₂ complex), Sigma-Aldrich®
Catalog # SIAL725455-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:1440.07 g/mol
Melting Pt:260-263 °C
:M-Clarity™ quality level = MQ100
:GHS08
CAS Number:142214-04-8
:68803-79-2
Formula:C₈₀H₆₀O₈Rh₂·CH₂Cl₂
MDL Number:MFCD15071082